Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CDH1 Rabbit Polyclonal Antibody

Supplier: Sino Biological

50671-RP01-400 50671-RP01-200 50671-RP01-100
103660-706EA 214.31 USD InStock
103660-706 103658-720 103658-718
Anti-CDH1 Rabbit Polyclonal Antibody
Antibodies
Anti-E-Cadherin Antibody

Type: Primary
Antigen: CDH1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty