Anti-CDH1 Rabbit Monoclonal Antibody [clone:117]

Supplier: Sino Biological
50671-R117-100 50671-R117-50
103595-428EA 150.18 USD
103595-428 103658-716
Anti-CDH1 Rabbit Monoclonal Antibody [clone:117]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse E-Cadherin/CDH1/E-cad/CD324 (rM E-Cadherin/CDH1/E-cad/CD324; Catalog#50671-M08H; P09803; Met1-Val709).

Type: Primary
Antigen: CDH1
Clonality: Monoclonal
Clone: 117
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR