Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IFNGR2 Rabbit Monoclonal Antibody [clone: 007]

Supplier: Sino Biological

50665-R007-50 50665-R007-100
103595-416EA 128.15 USD InStock
103595-416 103658-686
Anti-IFNGR2 Rabbit Monoclonal Antibody [clone: 007]
Antibodies
Anti-IFNGR2 Antibody

Type: Primary
Antigen: IFNGR2
Clonality: Monoclonal
Clone: Clone: 007
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty