Anti-CA9 Rabbit Monoclonal Antibody [clone: 001]

Supplier: Sino Biological

50660-R001-50 50660-R001-100
103595-406EA 128.15 USD
103595-406 103659-462
Anti-CA9 Rabbit Monoclonal Antibody [clone: 001]
Antibodies
Anti-Carbonic Anhydrase IX Ant

Type: Primary
Antigen: CA9
Clonality: Monoclonal
Clone: Clone: 001
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR