Anti-FABP4 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50652-RP02-100 50652-RP02-200 50652-RP02-50
103595-394EA 142.96 USD
103595-394 103658-666 103658-668
Anti-FABP4 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse FABP4 (rM FABP4; Catalog#50652-M08H; NP_077717.1; Met 1-Ala 132). FABP4 specific IgG was purified by mouse FABP4 affinity chromatography
.

Type: Primary
Antigen: FABP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR