Anti-GSK3B Rabbit Polyclonal Antibody

Supplier: Sino Biological
50650-T38-50 50650-T38-100 50650-T38-200
103595-392EA 352.03 USD
103595-392 103595-390 103589-226
Anti-GSK3B Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse GSK3B (rM GSK3B; Catalog#50650-M07B; Q9WV60; Met1-Thr420). GSK3B specific IgG was purified by Mouse GSK3B affinity chromatography.

Type: Primary
Antigen: GSK3B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR