Anti-CNTNAP2 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50649-RP02-200 50649-RP02-50 50649-RP02-100
103658-662EA 270.87 USD
103658-662 103658-664 103595-388
Anti-CNTNAP2 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse CNTNAP2 / CASPR2 (rM CNTNAP2 / CASPR2; Catalog#50649-M08H; NP_001004357.2; Met 1-Ser 1262). CNTNAP2 / CASPR2 specific IgG was purified by Mouse CNTNAP2 / CASPR2 affinity chromatography.

Type: Primary
Antigen: CASPR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR