Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CASPR2 Rabbit Polyclonal Antibody

Supplier: Sino Biological

50649-RP01-200 50649-RP01-400 50649-RP01-100
103658-658EA 160.46 USD InStock
103658-658 103658-660 103658-656
Anti-CASPR2 Rabbit Polyclonal Antibody
Antibodies
Anti-CASPR2 Antibody

Type: Primary
Antigen: CASPR2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty