Anti-CNTNAP2 Rabbit Monoclonal Antibody [clone:001]

Supplier: Sino Biological
50649-R001-100 50649-R001-50
103658-654EA 271.4 USD
103658-654 103595-386
Anti-CNTNAP2 Rabbit Monoclonal Antibody [clone:001]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CNTNAP2 / CASPR2 (rM CNTNAP2 / CASPR2; Catalog#50649-M08H; NP_001004357.2; Met 1-Ser 1262).

Type: Primary
Antigen: CASPR2
Clonality: Monoclonal
Clone: 001
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR