Anti-SEMA4D Rabbit Polyclonal Antibody

Supplier: Sino Biological

50642-RP02-50 50642-RP02-100 50642-RP02-200
103595-384EA 106.62 USD
103595-384 103658-650 103658-652
Anti-SEMA4D Rabbit Polyclonal Antibody
Antibodies
Anti-Semaphorin 4D Antibody

Type: Primary
Antigen: SEMA4D
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR