Anti-SEMA4D Rabbit Polyclonal Antibody

Supplier: Sino Biological
50642-RP01-100 50642-RP01-200 50642-RP01-400
103658-644EA 270.2 USD
103658-644 103658-648 103658-646
Anti-SEMA4D Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse Semaphorin 4D / SEMA4D / CD100 (rM Semaphorin 4D / SEMA4D / CD100; Catalog#50642-M08H; NP_038688.2; Met 1-Arg 733). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: SEMA4D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR