Anti-SEMA4D Rabbit Monoclonal Antibody (PE (Phycoerythrin)) [clone: 010]

Supplier: Sino Biological

50642-R010-P-100 50642-R010-P-25
103632-292EA 258.46 USD
103632-292 103630-128
Anti-SEMA4D Rabbit Monoclonal Antibody (PE (Phycoerythrin)) [clone: 010]
Antibodies
Anti-Semaphorin 4D Antibody (P

Type: Primary
Antigen: SEMA4D
Clonality: Monoclonal
Clone: Clone: 010
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR