Anti-SEMA4D Rabbit Monoclonal Antibody [clone: 010]

Supplier: Sino Biological

50642-R010-100 50642-R010-50
103658-640EA 214.31 USD
103658-640 103595-380
Anti-SEMA4D Rabbit Monoclonal Antibody [clone: 010]
Antibodies
Anti-Semaphorin 4D Antibody

Type: Primary
Antigen: SEMA4D
Clonality: Monoclonal
Clone: Clone: 010
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR