Anti-SEMA4D Rabbit Monoclonal Antibody [clone:006]

Supplier: Sino Biological
50642-R006-50 50642-R006-1 50642-R006-100
103589-222EA 1500.59 USD
103589-222 103658-638 103595-378
Anti-SEMA4D Rabbit Monoclonal Antibody [clone:006]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Semaphorin 4D / SEMA4D / CD100 (rM Semaphorin 4D / SEMA4D / CD100; Catalog#50642-M08H; NP_038688.2; Met1-Arg733).

Type: Primary
Antigen: SEMA4D
Clonality: Monoclonal
Clone: 006
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR