Anti-DLL4 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50640-RP01-400 50640-RP01-200 50640-RP01-100
103658-620EA 562.97 USD
103658-620 103658-616 103658-618
Anti-DLL4 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse DLL4 (rM DLL4; Catalog#50640-M08H; NP_062327.2; Met 1-Pro 525). Total IgG was purified by Protein A affinity chromatography
.

Type: Primary
Antigen: DLL4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR