Anti-DLL4 Rabbit Monoclonal Antibody [clone:004]

Supplier: Sino Biological
50640-R004-100 50640-R004-50 50640-R004-1
103595-372EA 150.18 USD
103595-372 103658-612 103589-218
Anti-DLL4 Rabbit Monoclonal Antibody [clone:004]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse DLL4 / Delta-like 4 (rM DLL4 / Delta-like 4; Catalog#50640-M08H; NP_062327.2; Met1-Pro525).

Type: Primary
Antigen: DLL4
Clonality: Monoclonal
Clone: 004
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR