Anti-CD200R4 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50639-RP01-100 50639-RP01-200 50639-RP01-400
103658-604EA 351.49 USD
103658-604 103658-602 103658-606
Anti-CD200R4 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse CD200R4 (rM CD200R4; Catalog#50639-M08H; NP_997127.1; Met 1-Thr 241). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: CD200R4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR