Anti-ADIPOQ Rabbit Polyclonal Antibody

Supplier: Sino Biological
50636-RP01-200 50636-RP01-400 50636-RP01-100
103658-596EA 351.49 USD
103658-596 103658-594 103659-446
Anti-ADIPOQ Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse ADIPOQ (rM ADIPOQ; Catalog#50636-M08H; NP_033735.3; Met 1-Asn 247). Total IgG was purified by Protein A affinity chromatography

Type: Primary
Antigen: ADIPOQ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR