Anti-ADIPOQ Rabbit Monoclonal Antibody [clone:038]

Supplier: Sino Biological
50636-R038-100 50636-R038-50
103595-360EA 150.18 USD
103595-360 103658-592
Anti-ADIPOQ Rabbit Monoclonal Antibody [clone:038]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Adiponectin / Acrp30 / ADIPOQ (rM Adiponectin / Acrp30 / ADIPOQ; Catalog#50636-M08H; NP_033735.3; Met 1-Asn 247).

Type: Primary
Antigen: ADIPOQ
Clonality: Monoclonal
Clone: 038
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR