Anti-EphA6 Rabbit Polyclonal Antibody

Supplier: Sino Biological

50630-T24-200 50630-T24-50 50630-T24-100
103595-358EA 214.31 USD
103595-358 103589-216 103595-356
Anti-EphA6 Rabbit Polyclonal Antibody
Antibodies
Anti-EphA6 Antibody

Type: Primary
Antigen: EphA6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR