Anti-EphA6 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50630-RP01-100 50630-RP01-200 50630-RP01-400
103658-586EA 562.97 USD
103658-586 103658-584 103658-582
Anti-EphA6 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse EphA6 (rM EphA6; Catalog#50630-M08H; NP_031964.2; Met 1-Gln 546). Total IgG was purified by Protein A affinity chromatography
.

Type: Primary
Antigen: EphA6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR