Anti-EphA6 Rabbit Monoclonal Antibody [clone:033]

Supplier: Sino Biological
50630-R033-50 50630-R033-100
103595-352EA 150.18 USD
103595-352 103658-580
Anti-EphA6 Rabbit Monoclonal Antibody [clone:033]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse EphA6 / EHK2 (rM EphA6 / EHK2; Catalog#50630-M08H; NP_031964.2; Met 1-Gln 546).

Type: Primary
Antigen: EphA6
Clonality: Monoclonal
Clone: 033
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR