Anti-SERPINB10 Rabbit Polyclonal Antibody

Supplier: Sino Biological

50626-T16-100 50626-T16-50 50626-T16-200
103595-348EA 160.46 USD
103595-348 103589-214 103595-350
Anti-SERPINB10 Rabbit Polyclonal Antibody
Antibodies
Anti-SERPINB10 Antibody

Type: Primary
Antigen: SERPINB10
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR