Anti-CES2E Rabbit Polyclonal Antibody

Supplier: Sino Biological
50620-T48-50 50620-T48-100 50620-T48-200
103595-324EA 270.87 USD
103595-324 103595-326 103589-208
Anti-CES2E Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse CES5 / Carboxylesterase 5 (rM CES5 / Carboxylesterase 5; Catalog#50620-M08H; NP_766347.1; Met1-His556). CES5 / Carboxylesterase 5 specific IgG was purified by Mouse CES5 / Carboxylesterase 5 affinity chromatography.

Type: Primary
Antigen: CES7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR