Anti-EFNB2 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50598-T24-50 50598-T24-200 50598-T24-100
103589-204EA 142.96 USD
103589-204 103594-598 103594-596
Anti-EFNB2 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse Ephrin-B2 (Catalog#50598-M08H; NP_034241.2; Met1-Ala232). Ephrin-B2 specific IgG was purified by Mouse Ephrin-B2 affinity chromatography.

Type: Primary
Antigen: EPHB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR