Anti-EFNA5 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50597-T08-100 50597-T08-200 50597-T08-50
103595-318EA 270.87 USD
103595-318 103589-202 103595-320
Anti-EFNA5 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse Ephrin-A5 / EFNA5 (rM Ephrin-A5 / EFNA5; Catalog#50597-M08H; O08543-1; Met1-Asn203). Ephrin-A5 / EFNA5 specific IgG was purified by Mouse Ephrin-A5 / EFNA5 affinity chromatography.

Type: Primary
Antigen: EFNA5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR