Anti-EFNA3 Rabbit Polyclonal Antibody

Supplier: Sino Biological

50594-T24-50 50594-T24-200 50594-T24-100
103589-198EA 106.62 USD
103589-198 103595-310 103595-308
Anti-EFNA3 Rabbit Polyclonal Antibody
Antibodies
Anti-Ephrin A3 Antibody

Type: Primary
Antigen: EFNA3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR