Anti-EFNA1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50593-RP01-100 50593-RP01-200 50593-RP01-400
103659-424EA 270.2 USD
103659-424 103659-426 103659-428
Anti-EFNA1 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse Ephrin-A1 / EFNA1 (rM Ephrin-A1 / EFNA1; Catalog#50593-M08H;NP_034237.3; Met 1-Ser 182). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: EPHA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR