Anti-EpCAM Rabbit Polyclonal Antibody

Supplier: Sino Biological
50591-RP02-100 50591-RP02-50 50591-RP02-200
103659-420EA 270.87 USD
103659-420 103659-422 103595-304
Anti-EpCAM Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse EpCAM / TROP-1 / TACSTD1 (rM EpCAM / TROP-1 / TACSTD1; Catalog#50543-M08H; NP_032558.2; Met1-Thr266). EpCAM / TROP-1 / TACSTD1 specific IgG was purified by Mouse Acetylcholinesterase / ACHE affinity chromatography.

Type: Primary
Antigen: EpCAM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR