Anti-EpCAM Rabbit Polyclonal Antibody

Supplier: Sino Biological
50591-RP01-400 50591-RP01-200 50591-RP01-100
103659-414EA 270.2 USD
103659-414 103659-416 103659-418
Anti-EpCAM Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse EpCAM / TROP-1 / TACSTD1 (rM EpCAM / TROP-1 / TACSTD1; Catalog#50591-M08H; NP_032558.2; Met 1-Thr 266). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: EpCAM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR