Anti-CD34 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50589-T24-50 50589-T24-200 50589-T24-100
103595-294EA 270.87 USD
103595-294 103595-296 103589-192
Anti-CD34 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse CD34 (rM CD34; Catalog#50589-M08H; NP_001104529.1; Met1-Thr287). CD34 specific IgG was purified by Mouse CD34 affinity chromatography.

Type: Primary
Antigen: CD34
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR