Anti-CD34 Rabbit Polyclonal Antibody

Supplier: Sino Biological

50589-RP01-400 50589-RP01-200 50589-RP01-100
103658-522EA 214.31 USD
103658-522 103658-520 103658-518
Anti-CD34 Rabbit Polyclonal Antibody
Antibodies
Anti-CD34 Antibody

Type: Primary
Antigen: CD34
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR