Anti-CD34 Rabbit Monoclonal Antibody [clone:028]

Supplier: Sino Biological
50589-R028-100 50589-R028-1 50589-R028-50
103658-514EA 271.4 USD
103658-514 103589-190 103595-288
Anti-CD34 Rabbit Monoclonal Antibody [clone:028]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD34 (rM CD34; Catalog#50589-M08H; NP_001104529.1; Met1-Thr287).

Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: 028
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR