Anti-CD34 Rabbit Monoclonal Antibody [clone:013] FITC (Fluorescein Isothiocyanate)

Supplier: Sino Biological
50589-R013-F-100 50589-R013-F-25
103630-120EA 70.7 USD
103630-120 103632-284
Anti-CD34 Rabbit Monoclonal Antibody [clone:013] FITC (Fluorescein Isothiocyanate)
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD34 (rM CD34; Catalog#50589-M08H; NP_001104529.1; Met 1-Thr 287) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.

Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: Clone: 013
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR