Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD34 (PerCP) Rabbit Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 013]

Supplier: Sino Biological

50589-R013-C-100 50589-R013-C-25
103632-282EA 204.62 USD InStock
103632-282 103630-118
Anti-CD34 (PerCP) Rabbit Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 013]
Antibodies
Anti-CD34 Antibody (PerCP)

Type: Primary
Antigen: CD34 (PerCP)
Clonality: Monoclonal
Clone: Clone: 013
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty