Anti-EphA7 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50587-RP01-200 50587-RP01-100 50587-RP01-400
103658-502EA 351.49 USD
103658-502 103658-506 103658-504
Anti-EphA7 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse EphA7 / Eph Receptor A7 (Catalog#50587-M08H; Q61772-1; Met1-Ile556). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: EphA7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR