Anti-EphA2 Rabbit Polyclonal Antibody

Supplier: Sino Biological

50586-RP02-50 50586-RP02-200 50586-RP02-100
103595-282EA 106.62 USD
103595-282 103659-412 103659-410
Anti-EphA2 Rabbit Polyclonal Antibody
Antibodies
Anti-EphA2 Antibody

Type: Primary
Antigen: EphA2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR