Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-EphA2 Rabbit Monoclonal Antibody [clone: 301]

Supplier: Sino Biological

50586-R301-50 50586-R301-100
103595-280EA 128.15 USD InStock
103595-280 103659-402
Anti-EphA2 Rabbit Monoclonal Antibody [clone: 301]
Antibodies
Anti-EphA2 Antibody

Type: Primary
Antigen: EphA2
Clonality: Monoclonal
Clone: Clone: 301
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty