Anti-EphA2 Rabbit Monoclonal Antibody [clone:101]

Supplier: Sino Biological
50586-R101-100 50586-R101-50
103659-400EA 271.4 USD
103659-400 103595-278
Anti-EphA2 Rabbit Monoclonal Antibody [clone:101]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse EphA2 / Eph Receptor A2 (rM EphA2 / Eph Receptor A2; Catalog#50586-M08H; NP_034269.2; Met1-Asn535).

Type: Primary
Antigen: EphA2
Clonality: Monoclonal
Clone: 101
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR