Anti-VTN Rabbit Monoclonal Antibody [clone:019]

Supplier: Sino Biological
50585-R019-50 50585-R019-100
103595-274EA 150.18 USD
103595-274 103658-492
Anti-VTN Rabbit Monoclonal Antibody [clone:019]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Vitronectin / VTN (rM Vitronectin / VTN; Catalog#50585-M08H; NP_035837.1; Met1-Lys478).

Type: Primary
Antigen: VTN
Clonality: Monoclonal
Clone: 019
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR