Anti-CEL Rabbit Polyclonal Antibody

Supplier: Sino Biological
50583-RP01-200 50583-RP01-400 50583-RP01-100
103658-484EA 562.97 USD
103658-484 103658-482 103658-480
Anti-CEL Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse CEL (rM CEL; Catalog#50583-M08B; NP_034015.1; Met1-Leu534). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: CEL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR