Anti-EphB3 Rabbit Monoclonal Antibody [clone:115]

Supplier: Sino Biological
50581-R115-50 50581-R115-100
103659-388EA 271.4 USD
103659-388 103595-256
Anti-EphB3 Rabbit Monoclonal Antibody [clone:115]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse EphB3 / HEK2 (rM EphB3 / HEK2; Catalog#50581-M08H; NP_034273.1; Met 1-Thr 537).

Type: Primary
Antigen: EphB3
Clonality: Monoclonal
Clone: 115
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR