Anti-EphB3 Rabbit Monoclonal Antibody [clone:001] PE (Phycoerythrin)

Supplier: Sino Biological
50581-R001-P-100 50581-R001-P-25
103630-104EA 111.1 USD
103630-104 103631-948
Anti-EphB3 Rabbit Monoclonal Antibody [clone:001] PE (Phycoerythrin)
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse EphB3 / HEK2 (rM EphB3 / HEK2; Catalog#50581-M08H; NP_034273.1; Met1-Thr537) and conjugated with PE under optimum conditions, the unreacted PE was removed.

Type: Primary
Antigen: EphB3
Clonality: Monoclonal
Clone: Clone: 001
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR