Anti-CHST3 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50577-T24-200 50577-T24-100 50577-T24-50
103589-182EA 142.96 USD
103589-182 103595-242 103595-244
Anti-CHST3 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse CHST3 / C6ST-1 (rM CHST3 / C6ST-1; Catalog#50577-M07H; NP_058083.2; Glu39-Thr472). CHST3 / C6ST-1 specific IgG was purified by Mouse CHST3 / C6ST-1 affinity chromatography.

Type: Primary
Antigen: CHST3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR