Anti-14-3-3 sigma Rabbit Polyclonal Antibody

Supplier: Sino Biological

50576-RP01-200 50576-RP01-100 50576-RP01-400
103658-446EA 160.46 USD
103658-446 103658-444 103658-448
Anti-14-3-3 sigma Rabbit Polyclonal Antibody
Antibodies
Anti-14-3-3 sigma Antibody

Type: Primary
Antigen: 14-3-3 sigma
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR