Anti-FOLR1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50573-RP02-100 50573-RP02-200 50573-RP02-50
103658-442EA 352.03 USD
103658-442 103658-440 103595-234
Anti-FOLR1 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse FOLR1 (rM FOLR1; Catalog#50573-M08H; NP_032060.2; Met 1-Met 231). FOLR1 specific IgG was purified by Mouse FOLR1 affinity chromatography.

Type: Primary
Antigen: FOLR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR