Anti-CEACAM1 Rabbit Monoclonal Antibody [clone:117] PE (Phycoerythrin)

Supplier: Sino Biological
50571-R117-P-100 50571-R117-P-25
103631-944EA 222.2 USD
103631-944 103630-100
Anti-CEACAM1 Rabbit Monoclonal Antibody [clone:117] PE (Phycoerythrin)
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CEACAM1 / CD66a (rM CEACAM1 / CD66a; Catalog#50571-M08H; NP_001034274.1; Met1-Gly428) and conjugated with PE under optimum conditions, the unreacted PE was removed.

Type: Primary
Antigen: CEACAM1
Clonality: Monoclonal
Clone: Clone: 117
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR