Anti-CEACAM1 Rabbit Monoclonal Antibody [clone: 117]

Supplier: Sino Biological

50571-R117-100 50571-R117-50
103658-408EA 214.31 USD
103658-408 103595-224
Anti-CEACAM1 Rabbit Monoclonal Antibody [clone: 117]
Antibodies
Anti-CEACAM1 Antibody

Type: Primary
Antigen: CEACAM1
Clonality: Monoclonal
Clone: Clone: 117
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR