Anti-CEACAM1 Rabbit Monoclonal Antibody [clone:025]

Supplier: Sino Biological
50571-R025-50 50571-R025-100
103658-404EA 271.4 USD
103658-404 103595-220
Anti-CEACAM1 Rabbit Monoclonal Antibody [clone:025]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CEACAM1 / CD66a (rM CEACAM1 / CD66a; Catalog#50571-M08H; NP_001034274.1; Met1-Gly428).

Type: Primary
Antigen: CEACAM1
Clonality: Monoclonal
Clone: 025
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR