Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CDNF Rabbit Polyclonal Antibody

Supplier: Sino Biological

50569-RP02-200 50569-RP02-100 50569-RP02-50
103658-402EA 214.31 USD InStock
103658-402 103658-400 103595-214
Anti-CDNF Rabbit Polyclonal Antibody
Antibodies
Anti-CDNF Antibody

Type: Primary
Antigen: CDNF
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty